Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Acetic Acid Glacial
SDP

Catalog No. 21033256
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-033-256 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-256 Supplier StatLab Supplier No. 0010016
Add to Cart
Add to Cart

Reagent ACS grade glacial acetic acid

  • Used as a component in fixative solutions
  • Helps to preserve tissue structure by preventing shrinkage and maintaining the integrity of cell nuclei
  • Enhances the preservation of chromosomes during cytogenetic examinations and contributes to clearer cellular details in slides

Specifications

Capacity 16 oz.
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.