Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, 2.5 L, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S000999 Shop All Aldon Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000999 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000999 Supplier Aldon Corporation Supplier No. IS10010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-19-7
Molecular Weight (g/mol) 60.05

Specifications

Quantity 2.5 L
Packaging Glass
Percent Purity 99.80%
Grade Laboratory
Chemical Name or Material Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.