Learn More
Research Products International Corp Acetic Acid, Glacial, 2.5 Liters
SDP
Catalog No. 50712720
Encompass_Preferred
Acetic acid (CH₃COOH) is a colorless, corrosive liquid with a strong odor, fully miscible in water. With a molecular weight of 60.05 g/mol, it is used in life science labs for buffer preparation, protein precipitation, histological staining, pH regulation, and as a solvent in various biochemical and molecular procedures.
Catalog No. 50-712-720 Supplier Research Products International Corp Supplier No. A128002500.0
Acetic acid (CH₃COOH) is a colorless, corrosive liquid with a strong odor, fully miscible in water. With a molecular weight of 60.05 g/mol, it is used in life science labs for buffer preparation, protein precipitation, histological staining, pH regulation, and as a solvent in various biochemical and molecular procedures.