Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chem-Impex International, Inc. Acetic acid, glacial | MFCD00036152 | 4L

Catalog No. NC2020330
Encompass_Preferred

ACETIC ACID, GLACIAL 4L

Catalog No. NC2020330 Supplier Chem-Impex International, Inc. Supplier No. 301734L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACETIC ACID, GLACIAL 4L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.