Learn More
Acetic acid, glacial, ACS, 99.7+%
Description
It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 16.6°C |
| Boiling Point | 118.1°C |
| Assay Percent Range | 99.7+% |
| UN Number | UN2789 |
| Merck Index | 14,55 |
| Solubility Information | Miscible with water. |
| Formula Weight | 60.05 |
| Percent Purity | ≥99.7% |
| Grade | ACS |
| Density | 1.049 |
| Show More |
Safety and Handling
| Hazard Category | H226-H314 |
| Hazard Statement | GHS H Statement H314-H226 |
| Precautionary Statement | P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL |
| EINECSNumber | 200-580-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.