Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RABA0010250 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Glass Bottle
Hard Pack
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABA0010250 250 mL Natural Poly Bottle
RABA0010500 500 mL Glass Bottle
RABA00101C 1 L Amber Glass Bottle
RABA001025D 2.5 L Glass Bottle
RABA00104C 4 L Amber Glass Bottle
RABA001020E 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RABA0010250 Supplier Ricca Chemical Company Supplier No. RABA0010250A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 98.0
Color Colorless
CAS Max % 100.0
Synonym ethanoic acid
Percent Purity 99.7% (min.)
Grade ACS
Quantity 250 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Residue after Evaporation max. 0.001%
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.