Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial (Certified ACS Plus), Fisher BioReagents

Catalog No. BP2401SI212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 2.5 L
Packaging:
Glass
Safe-Cote™ Glass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2401SI212 2.5 L Safe-Cote™ Glass
BP2401-500 500 mL Glass
BP2401-212 2.5 L Glass
BP2401C-212 6 x 2.5 L Glass
BP2401S-212 6 x 2.5 L Safe-Cote™ Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BP2401SI212
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Acetic Acid, Glacial can be used for several biochemical procedures. Some products are packed in Safe Cote™ which give additional safety for chemical and user both.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Density 1.048g/cm³
Quantity 2.5 L
Packaging Safe-Cote™ Glass
Physical Form Liquid
Residue after Evaporation 8ppm max.
Linear Formula CH3COOH
Vapor Pressure 1.52 kPa at 20°C
Formula Weight 60.05
Percent Purity ≥99.7%
Grade Certified ACS Plus
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.