Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, HPLC Grade, J.T. Baker™

Catalog No. 02002123 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Clear Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-123 2.5 L Clear Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-123 Supplier Avantor Supplier No. 951503
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity solvents for use in analytical and preparative separations

    For Use in High Performance Liquid Chromatography, Bulk Pharmaceutical Chemical

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Quantity 2.5 L
Flash Point 39.00°C
Packaging Clear Glass Bottle
Specific Gravity 1L = 1.05kg
Formula Weight 60.05
Grade Meets USP requirements
Chemical Name or Material Acetic Acid, Glacial
Recommended Storage IMPORTANT: Material will freeze if stored below 17°C (63°F), Preserve in tight containers, and store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.