Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetic acid glacial puriss
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 272252.5LR
Encompass
Acetic acid is an organic carboxylic acid. It is a hygroscopic liquid having pungent odor. Its anhydrous form is commonly referred as glacial acetic acid. It forms dimers linked by hydrogen bonds in SOLUTIONon and gaseous phases. It is weakly acidic in nature.
Catalog No. 50-289-8649