Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetic acid, phenyl ester | 122-79-2 | 99.7% | 5 G
SDP

Catalog No. 5000339353
Encompass_Preferred

Phenyl acetate is an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. It regulates glutamine metabolism and can serve as a detection index for conditions like depression. This product is intended for research use only.

  • Endogenous metabolite
  • Metabolite of 2-phenylethylamine
  • Regulates glutamine metabolism
  • Can serve as a detection index for conditions like depression
  • Purity: 99.68%
  • Molecular weight: 136.15
  • Appearance: Colorless to light yellow liquid
  • Can block glutaminolysis in lung cancer cell lines

Catalog No. 50-003-39353 Supplier Medchemexpress LLC Supplier No. HY1287335G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenyl acetate is an endogenous metabolite and a metabolite of 2-phenylethylamine, found in urine. It regulates glutamine metabolism and can serve as a detection index for conditions like depression. This product is intended for research use only.

  • Endogenous metabolite
  • Metabolite of 2-phenylethylamine
  • Regulates glutamine metabolism
  • Can serve as a detection index for conditions like depression
  • Purity: 99.68%
  • Molecular weight: 136.15
  • Appearance: Colorless to light yellow liquid
  • Can block glutaminolysis in lung cancer cell lines

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.