Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid Solution, 40mL/gallon, Reagents

Catalog No. 01335913
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-913 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-913 Supplier Reagents Holdings Llc Supplier No. CA2037004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetic Acid Solution, 40mL/gallon, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetic Acid Solution, 40mL/gallon
pH 4.5
CAS 7732-18-5, 64-19-7
Boiling Point 100°C
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0005
Molecular Formula CH3COOH/H2O
Synonym Ethanoic Acid, Ethylic Acid
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.