Learn More
Acetic Acid/Sulfuric Acid Solution, Reagents
Specifications
Specifications
| Chemical Name or Material | Acetic Acid/Sulfuric Acid Solution |
| CAS | 64-19-7, 7732-18-5, 7732-18-5 |
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1.08 |
| Synonym | Ethanoic Acid/Sulphuric Acid |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.