Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, TraceSELECT™, ≥99%, Solstice

Catalog No. 60046914 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-914 500 mL Poly Bottle
60-046-912 1 L Poly Bottle
60-046-913 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-914 Supplier Solstice Supplier No. 45727500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, for trace analysis,≥99.0%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.2°C
Boiling Point 117°C to 118°C
Color Colorless
Appearance Liquid
Density 1.049g/mL at 25°C
Quantity 500 mL
Explosion Limit 4% at 59°F, 16% at 92°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Poly Bottle
Refractive Index n20/D 1.371
Linear Formula CH3CO2H
Vapor Density 2.07 (vs air)
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99%
Grade TraceSELECT
Chemical Name or Material Acetic acid
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P310
DOTInformation 3(8)/PGII
RTECSNumber 200-580-7
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.