Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial (Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. A38212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
6 x 2.5 L
450 lb.
Packaging:
Glass Bottle
Poly Drum
Safe-Cote™ Glass
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A38-212 2.5 L Glass Bottle
A38-500 500 mL Glass Bottle
A38S-500 500 mL Safe-Cote™ Glass
A38S-212 2.5 L Safe-Cote™ Glass
A38C-212 6 x 2.5 L Glass Bottle
A38P-20 20 L Poly Drum
A38-450LB 450 lb. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. A38-212
Only null left
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Assay Percent Range Assay: ≥99.7% w/w
Linear Formula CH3COOH
UN Number 2789
Merck Index 15, 54
Solubility Information Soluble in water
Formula Weight 60.05
Percent Purity ≥99.7% (w/w)
Grade Certified ACS
Density 1.048g/cm³
Quantity 2.5 L
Flash Point 40°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 8ppm max.
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 8: Corrosive
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.