Learn More
Cambridge Isotope Laboratories ACETIC ANHYDRIDE (1,1',2,2'-13C4, 99%), 1 G, 114510-14-4
Supplier:  Cambridge Isotope Laboratories CLM11611
Encompass
ACETIC ANHYDRIDE (1,1',2,2'-13C4, 99%), 1 G, 114510-14-4
Catalog No. 50-163-0060