Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetic Anhydride
SCI_ED

Catalog No. S25119A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Lab Grade
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25119A 500 mL Reagent Grade
S25119 100 mL Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25119A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Density 1.0820g/cmsuperscript_3;
Assay Percent Range >97% w/v
Packaging Reagent Grade
Linear Formula (CH3CO)2O
Quantity 500 mL
Formula Weight 102.09
Chemical Name or Material Acetic Anhydride
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 8 (3), II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.