Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic anhydride, ACS, 97+%

Catalog No. AA36292AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36292AP 500 mL
AA36292AE 100 mL
AA36292K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36292AP Supplier Thermo Scientific Chemicals Supplier No. 036292AP
Add to Cart
Edge

missing translation for 'validMainframeMsg'

CAS: 108-24-7 | C4H6O3 | 102.089 g/mol

It is most suited for the conversion of cellulose into cellulose acetate and in the manufacture of aspirin. It acts as a wood preservative and solvent in examination of wood fat, fatty and volatile oils and detection of rosins. In many organic reactions it acts as a dehydrating agent. Also used in the production of isoamyl alcohol and 2-methyl furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Melting Point °C to 73°C
Density 1.08
Boiling Point 138°C to 140°C
Flash Point 49°C (120°F)
Assay Percent Range 97+%
Refractive Index 1.39
Linear Formula (CH3CO)2O
Quantity 500 mL
UN Number UN1715
Beilstein 385737
Sensitivity Moisture sensitive
Merck Index 14,56
Solubility Information Soluble in ether,chloroform and benzene. Slowly soluble in water.
Formula Weight 102.09
Percent Purity ≥97%
Grade ACS
Physical Form Liquid
Chemical Name or Material Acetic anhydride
Show More Show Less
Hazard Category H226-H302-H314-H330
Hazard Statement GHS H Statement
H331-H314-H226-H302
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ANHYDRIDE
EINECSNumber 203-564-8
RTECSNumber AK1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.