Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride, ACS, MilliporeSigma™

Catalog No. MAX00806 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX00806 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX00806 Supplier MilliporeSigma Supplier No. AX00806
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For General Chemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

pH 3 (10g/L, H2O, 20°C)
Flash Point 49°C
Packaging Glass Bottle
Quantity 500 mL
Vapor Pressure 4 hPa (20°C)
Formula Weight 102.09
Percent Purity 99.0% min
Grade ACS
Physical Form Liquid
EINECSNumber 203-564-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.