Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride, BioSyn™, for DNA/RNA Synthesis, >99.5%, Solstice

Catalog No. 60048319 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-319 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-319 Supplier Solstice Supplier No. BR6494
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity acetic anhydride for use in laboratory chemical synthesis.

Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis

  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym Ethanoic Anhydride
IUPAC Name acetyl acetate
SMILES CC(=O)OC(C)=O

Specifications

Density 1.083 g/cm3
Boiling Point 140°C
Flash Point 49.5°C
Packaging Glass Bottle
Quantity 4 x 4 L
Beilstein 385737
Grade Capping Reagents
Physical Form Liquid
Hazard Category Danger
Hazard Statement H335+H314+H302+H332+H225
Precautionary Statement P210+P233+P240+P241+P242+P243+P261+P280+P271+P270+P264
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.