Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride (Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. A101 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A10-1 1 L Glass Bottle
A10-500 500 mL Glass Bottle
A10-100 100 mL Glass Bottle
A10-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A10-1
Only null left
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Melting Point -73.1°C
Color Colorless
Density 1.087g/cm³
pH 3
Boiling Point 140°C
Flash Point 53°C
Assay Percent Range ≥97.0 %
Packaging Glass Bottle
Linear Formula (CH3CO)2O
Quantity 1 L
UN Number 1715
Residue after Evaporation 0.002% max.
Formula Weight 102.09
Percent Purity ≥97.0%
Grade Certified ACS
Physical Form Liquid
Chemical Name or Material Acetic Anhydride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.