Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride, MP Biomedicals™

Catalog No. ICN15468090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15468090 500 mL
ICN15468091 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15468090 Supplier MP Biomedicals, Inc Supplier No. 0215468090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetic Anhydride is a reagent that is used in the manufacturing of acetyl compounds and cellulose acetates.

  • It is also utilized as a solvent in the examination of wool fat, glycerol, fatty and volatile oils, and resins.
  • In the synthesis of organic compounds, acetic anhydride is used in various reactions, including the cleavage of dialkyl ethers, the formation of anhydrides from carboxylic acids, the addition of methylthiomethyl groups to phenol rings, and the addition of acyl and acyloxy groups across double bonds.
  • Acetic anhydride has been used to hyperacetylate histones to investigate transcriptional effects of the NAD-dependent histone deacetylase Sir2 in Drosophila on a complex of the hyperacetylated histones with DNA.
  • Acetic anhydride is utilized in an assay of a fluorescently labeled peptide catalyst library for application to the resolution of secondary alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Color Colorless
Assay Percent Range 0.998
Quantity 500 mL
Formula Weight 102.09
Physical Form Liquid
Chemical Name or Material Acetic Anhydride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.