Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Aceto-Orcein Solution, 2%
SCI_ED

Catalog No. S25121 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
30 mL
100 mL
Packaging:
2%
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25121 30 mL 2%
S25121A 100 mL 2%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25121

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-19-7 , 1400-62-0 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym 2% Solution
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Assay Percent Range 31.5% v/v
Packaging 2%
Density 1.049
Color Violet
Quantity 30 mL
Chemical Name or Material Aceto-Orcein Solution, 2%
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.