Learn More
Medchemexpress LLC Acetoacetic acid | 541-50-4 | 95.0% | 102.09 | 1 MG
SDP
Catalog No. 5000436309
Encompass_Preferred
Acetoacetic acid is an oxidative stress inducer. It affects the antioxidant enzyme system and lipoprotein metabolism by decreasing mRNA expression and activity of superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GSH-Px), and increasing malondialdehyde (MDA) content. It also inhibits very low-density lipoprotein (VLDL) assembly, leading to triglyceride (TG) accumulation in hepatocytes, making it useful for studying metabolic diseases.
  • Oxidative stress inducer
  • Impacts antioxidant enzyme system
  • Affects lipoprotein metabolism
  • Decreases SOD, CAT, and GSH-Px activity and mRNA expression
  • Increases malondialdehyde (MDA) content
  • Inhibits VLDL assembly
  • Leads to triglyceride (TG) accumulation in hepatocytes
  • Useful for studying metabolic diseases
Catalog No. 50-004-36309 Supplier Medchemexpress LLC Supplier No. HY1125401MG
Acetoacetic acid is an oxidative stress inducer. It affects the antioxidant enzyme system and lipoprotein metabolism by decreasing mRNA expression and activity of superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GSH-Px), and increasing malondialdehyde (MDA) content. It also inhibits very low-density lipoprotein (VLDL) assembly, leading to triglyceride (TG) accumulation in hepatocytes, making it useful for studying metabolic diseases.
  • Oxidative stress inducer
  • Impacts antioxidant enzyme system
  • Affects lipoprotein metabolism
  • Decreases SOD, CAT, and GSH-Px activity and mRNA expression
  • Increases malondialdehyde (MDA) content
  • Inhibits VLDL assembly
  • Leads to triglyceride (TG) accumulation in hepatocytes
  • Useful for studying metabolic diseases