Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetoacetic acid | 541-50-4 | MFCD00053827 | 95.0% | 102.09 g/mol | C4H6O3 | 5 MG
SDP

Catalog No. 5000264317
Encompass_Preferred

Acetoacetic acid is a small-molecule research reagent used to induce oxidative stress and probe lipid metabolism in cellular models. It modulates antioxidant enzyme expression and activity and is used in studies of metabolic disease and hepatocyte triglyceride accumulation.

  • Acts as an oxidative stress inducer in cellular assays.
  • Affects antioxidant enzymes such as SOD, CAT, and GSH-Px.
  • Useful for studying lipid metabolism and triglyceride accumulation.
  • Purity 95.0% for research applications.
  • Chemical formula C4H6O3; molecular weight 102.09 g/mol.
  • Available in small milligram pack sizes for experimental use.

Catalog No. 50-002-64317 Supplier Medchemexpress LLC Supplier No. HY1125405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetoacetic acid is a small-molecule research reagent used to induce oxidative stress and probe lipid metabolism in cellular models. It modulates antioxidant enzyme expression and activity and is used in studies of metabolic disease and hepatocyte triglyceride accumulation.

  • Acts as an oxidative stress inducer in cellular assays.
  • Affects antioxidant enzymes such as SOD, CAT, and GSH-Px.
  • Useful for studying lipid metabolism and triglyceride accumulation.
  • Purity 95.0% for research applications.
  • Chemical formula C4H6O3; molecular weight 102.09 g/mol.
  • Available in small milligram pack sizes for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.