Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetoacetic acid (lithium) | 3483-11-2 | 98.0% | 108.02 | 100 MG
SDP

Catalog No. 5000436342
Encompass_Preferred

Acetoacetic acid (lithium) | 3483-11-2 | 98.0% | 108.02 | 100 MG

Catalog No. 50-004-36342 Supplier Medchemexpress LLC Supplier No. HY112540A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetoacetic acid (lithium) | 3483-11-2 | 98.0% | 108.02 | 100 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.