Learn More
Medchemexpress LLC Acetoacetic acid lithium | 3483-11-2 | 98.0% | 108.02 | 25 MG
SDP
Catalog No. 5000438926
Encompass_Preferred
Acetoacetic acid lithium is an oxidative stress inducer that impacts the antioxidant enzyme system and lipoprotein metabolism. It induces oxidative stress by decreasing the mRNA expression and activity of superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GSH-Px), increasing malondialdehyde (MDA) content. It also inhibits very low-density lipoprotein (VLDL) assembly by downregulating apolipoprotein ApoB100, ApoE, and low-density lipoprotein receptor (LDLR), leading to triglyceride (TG) accumulation in hepatocytes. This compound is useful for studying metabolic diseases.
  • Oxidative stress inducer
  • Affects the antioxidant enzyme system and lipoprotein metabolism
  • Decreases SOD, CAT, and GSH-Px expression and activity
  • Increases malondialdehyde (MDA) content
  • Inhibits VLDL assembly
  • Leads to triglyceride accumulation in hepatocytes
Catalog No. 50-004-38926 Supplier Medchemexpress LLC Supplier No. 25MGHY112540A
Acetoacetic acid lithium is an oxidative stress inducer that impacts the antioxidant enzyme system and lipoprotein metabolism. It induces oxidative stress by decreasing the mRNA expression and activity of superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GSH-Px), increasing malondialdehyde (MDA) content. It also inhibits very low-density lipoprotein (VLDL) assembly by downregulating apolipoprotein ApoB100, ApoE, and low-density lipoprotein receptor (LDLR), leading to triglyceride (TG) accumulation in hepatocytes. This compound is useful for studying metabolic diseases.
  • Oxidative stress inducer
  • Affects the antioxidant enzyme system and lipoprotein metabolism
  • Decreases SOD, CAT, and GSH-Px expression and activity
  • Increases malondialdehyde (MDA) content
  • Inhibits VLDL assembly
  • Leads to triglyceride accumulation in hepatocytes