Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acetoacetic acid (lithium) | 578486 | MFCD00013060 | 98.0% | 108.02 g/mol | C4H6LiO3 | 5 MG
SDP

Catalog No. 5000264322
Encompass_Preferred

Acetoacetic acid (lithium) is the lithium salt of acetoacetic acid supplied as a research reagent for biochemical and cell-based studies. It functions as an oxidative stress inducer and is provided as a small-quantity solid suitable for preparing aqueous or DMSO solutions for in vitro experiments.

  • Oxidative stress inducer for cell and biochemical assays.
  • High nominal purity (98.0%).
  • Supplied as a 5 mg vial for small-scale research use.
  • Soluble in water and DMSO for flexible formulation.
  • Molecular weight 108.02 g/mol; formula C4H6LiO3.
  • CAS number 3483-11-2 for unambiguous identification.

Catalog No. 50-002-64322 Supplier Medchemexpress LLC Supplier No. HY112540A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acetoacetic acid (lithium) is the lithium salt of acetoacetic acid supplied as a research reagent for biochemical and cell-based studies. It functions as an oxidative stress inducer and is provided as a small-quantity solid suitable for preparing aqueous or DMSO solutions for in vitro experiments.

  • Oxidative stress inducer for cell and biochemical assays.
  • High nominal purity (98.0%).
  • Supplied as a 5 mg vial for small-scale research use.
  • Soluble in water and DMSO for flexible formulation.
  • Molecular weight 108.02 g/mol; formula C4H6LiO3.
  • CAS number 3483-11-2 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.