Learn More
TARGETMOL CHEMICALS INC Acetoacetic acid lithium salt
SDP
Catalog No. 502879980
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Acetoacetic acid lithium salt (Lithium 3-Oxobutyrate) is a weak organic acid that can be produced in the human liver under certain conditions of poor Metabolism leading to excessive fatty acid breakdown (diabetes mellitus leading to diabetic ketoacidosis).
Catalog No. 50-287-9980 Supplier TARGETMOL CHEMICALS INC Supplier No. T485450MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Acetoacetic acid lithium salt (Lithium 3-Oxobutyrate) is a weak organic acid that can be produced in the human liver under certain conditions of poor Metabolism leading to excessive fatty acid breakdown (diabetes mellitus leading to diabetic ketoacidosis).