Learn More
Medchemexpress LLC HY-112540B 10mg , Acetoacetic acid sodium salt CAS:623-58-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112540B10MG
Encompass_Preferred
HY-112540B 10mg Acetoacetic acid sodium salt CAS: 623-58-5 Acetoacetic acid sodium salt is a metabolite of non-esterified fatty acids, involved in the development of human diabetes. Acetoacetic acid sodium salt induces oxidative stress to inhibit the assembly of very low density lipoprotein in bovine hepatocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9633