Learn More
Medchemexpress LLC Acetoacetyl-CoA sodium hydrate | 1420-36-6 | 1 MG
SDP
Catalog No. 5000418497
Encompass_Preferred
Acetoacetyl-CoA sodium hydrate is a crucial precursor of HMG-CoA in the mevalonate pathway and is essential for cholesterol biosynthesis. It is formed through the catalysis of two acetyl-CoA molecules by acetoacetyl-CoA thiolase, acting as an intermediate in the biological breakdown and synthesis of fatty acids.
  • Precursor of HMG-CoA in the mevalonate pathway.
  • Essential for cholesterol biosynthesis.
  • Intermediate in the biological breakdown and synthesis of fatty acids.
  • Formed by the enzyme acetoacetyl-CoA thiolase.
Catalog No. 50-004-18497 Supplier Medchemexpress LLC Supplier No. HYN7392A1MG
Acetoacetyl-CoA sodium hydrate is a crucial precursor of HMG-CoA in the mevalonate pathway and is essential for cholesterol biosynthesis. It is formed through the catalysis of two acetyl-CoA molecules by acetoacetyl-CoA thiolase, acting as an intermediate in the biological breakdown and synthesis of fatty acids.
  • Precursor of HMG-CoA in the mevalonate pathway.
  • Essential for cholesterol biosynthesis.
  • Intermediate in the biological breakdown and synthesis of fatty acids.
  • Formed by the enzyme acetoacetyl-CoA thiolase.