Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoacetyl coenzyme A sodium salt, >90%, MP Biomedicals™

Catalog No. ICN15022450 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15022450 50 mg
ICN15022425 25 mg
ICN15022401 1 mg
ICN15022410 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15022450 Supplier MP Biomedicals, Inc Supplier No. 0215022450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102029-52-7
Molecular Formula C25H40N7NaO18P3S
Molecular Weight (g/mol) 874.597
InChI Key RSOOKMDOLPTBRH-UHFFFAOYSA-N
PubChem CID 133112705
IUPAC Name S-[2-[3-[[4-[[[5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 3-oxobutanethioate;sodium
SMILES CC(=O)CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O.[Na]

Specifications

Color Yellow
Assay Percent Range >90%
Quantity 50 mg
Formula Weight 901.6 (free acid)
Physical Form Solid
Chemical Name or Material Acetoacetyl coenzyme A
Recommended Storage Store at -20°C.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.