Learn More
Medchemexpress LLC Acetobromo-α-D-galactose | 3068-32-4 | MFCD00063686 | ≥97.0% | 411.20 | C14H19BrO9 | 50 G
SDP
Catalog No. 5000301716
Encompass_Preferred
Acetobromo-α-D-galactose is a brominating agent used as an intermediate in the synthesis of glycosides and oligosaccharides. Supplied for research use only, it is provided as a white to off-white solid with reported purity ≥97.0% and molecular weight 411.20 g·mol-1.
  • Brominating agent for glycosylation and derivatization.
  • High reported purity of ≥97.0%.
  • White to off-white solid form.
  • Available in multiple lab-scale sizes (5 g-100 g, larger by quote).
  • Datasheet, COA, and SDS available.
Catalog No. 50-003-01716 Supplier Medchemexpress LLC Supplier No. HYW02881350G
Add to Cart
Add to Cart
Acetobromo-α-D-galactose is a brominating agent used as an intermediate in the synthesis of glycosides and oligosaccharides. Supplied for research use only, it is provided as a white to off-white solid with reported purity ≥97.0% and molecular weight 411.20 g·mol-1.
  • Brominating agent for glycosylation and derivatization.
  • High reported purity of ≥97.0%.
  • White to off-white solid form.
  • Available in multiple lab-scale sizes (5 g-100 g, larger by quote).
  • Datasheet, COA, and SDS available.