Learn More
Medchemexpress LLC Acetobromo-α-D-galactose | 3068-32-4 | MFCD00063686 | >97.0% | 411.20 g·mol⁻¹ | C14H19BrO9 | 100 G
SDP
Catalog No. 5000300173
Encompass_Preferred
Acetobromo-α-D-galactose is an acetylated glycosyl bromide used as a brominating reagent and intermediate in the chemical synthesis of glycosides and oligosaccharides. It is supplied as a solid with typical purity ≥97.0%, molecular formula C14H19BrO9, and molecular weight 411.20 g·mol⁻¹. Datasheet, COA, and SDS are available for characterization and safe handling.
  • Brominating reagent for glycosylation and carbohydrate synthesis.
  • Typical purity ≥97.0% supports reliable reactions.
  • Solid form simplifies storage and handling.
  • Documented analytical data available (datasheet, COA, SDS).
  • Characterized molecular formula C14H19BrO9 and molecular weight 411.20 g·mol⁻¹.
Catalog No. 50-003-00173 Supplier Medchemexpress LLC Supplier No. HYW028813100G
Acetobromo-α-D-galactose is an acetylated glycosyl bromide used as a brominating reagent and intermediate in the chemical synthesis of glycosides and oligosaccharides. It is supplied as a solid with typical purity ≥97.0%, molecular formula C14H19BrO9, and molecular weight 411.20 g·mol⁻¹. Datasheet, COA, and SDS are available for characterization and safe handling.
  • Brominating reagent for glycosylation and carbohydrate synthesis.
  • Typical purity ≥97.0% supports reliable reactions.
  • Solid form simplifies storage and handling.
  • Documented analytical data available (datasheet, COA, SDS).
  • Characterized molecular formula C14H19BrO9 and molecular weight 411.20 g·mol⁻¹.