Learn More
Medchemexpress LLC Acetobromo-α-D-glucuronic acid methyl ester | 25 G
SDP
Catalog No. 5000448142
Encompass_Preferred
Acetobromo-α-D-glucuronic acid methyl ester is a glucuronosyl donor that is applicable to the synthesis of other compounds.
  • Is a glucuronosyl donor
  • Applicable to the synthesis of other compounds
Catalog No. 50-004-48142 Supplier Medchemexpress LLC Supplier No. 25GHYW008125
Acetobromo-α-D-glucuronic acid methyl ester is a glucuronosyl donor that is applicable to the synthesis of other compounds.
  • Is a glucuronosyl donor
  • Applicable to the synthesis of other compounds