Learn More
Cayman Chemical AcetohexamIde 50mg
SDP
Supplier:  Cayman Chemical 2390050 MG
Encompass_Preferred
A first generation sulfonylurea; inhibits SUR1-KIR6.2 (Kis = 22.9 and 14.2 µM, in HEK293 cells and rat brain, respectively); metabolized to the hypoglycemic compound L-hydroxyhexamide in vivo
Catalog No. 50-296-4510