Learn More
Selleck Chemical LLC Acetohydroxamic acid S4602-100mg
SDP
Catalog No. 503112123
Encompass_Preferred
Acetohydroxamic acid is an Urease Inhibitor In the urine it acts as an antagonist of the bacterial enzyme urease
Catalog No. 50-311-2123 Supplier Selleck Chemical LLC Supplier No. S4602100MG
Acetohydroxamic acid is an Urease Inhibitor In the urine it acts as an antagonist of the bacterial enzyme urease