Learn More
Cayman Chemical Acetohydroxamic Acid, Purity: ≥98%, CAS: 546-88-3, 25 g
SDP
Catalog No. 502933092
Encompass_Preferred
An irreversible urease inhibitor; inhibits the growth of struvite crystals in artificial urine; inhibits the growth of H. pylori in vitro (MICs = 200 and 400 mg/L for various isolates); chronic administration reduces urine urease activity, pH, and crystalluria and inhibits growth of bladder stones in dogs; decreases gastritis, gastric lesions, and bacterial infection rats in gerbils (2,500 ppm/animal) following , H. pylori infection
Catalog No. 50-293-3092 Supplier Cayman Chemical Supplier No. 2397925 G
An irreversible urease inhibitor; inhibits the growth of struvite crystals in artificial urine; inhibits the growth of H. pylori in vitro (MICs = 200 and 400 mg/L for various isolates); chronic administration reduces urine urease activity, pH, and crystalluria and inhibits growth of bladder stones in dogs; decreases gastritis, gastric lesions, and bacterial infection rats in gerbils (2,500 ppm/animal) following , H. pylori infection