Learn More
Medchemexpress LLC Acetohydroxamic acid | 546-88-3 | 98.7% | 1 ML
SDP
Catalog No. 5000412906
Encompass_Preferred
Acetohydroxamic acid is an inhibitor for bacterial and plant urease used for chronic urinary tract infections. It selectively inhibits arachidonic acid 5-lipoxygenase, useful in asthma research. This compound also inhibits advanced glycation end products, reduces oxidative stress and inflammatory responses, and exhibits antiviral activity against HIV.
  • Inhibitor for bacterial and plant urease.
  • Selectively inhibits arachidonic acid 5-lipoxygenase, applicable in asthma research.
  • Inhibits the formation of advanced glycation end products (AGEs).
  • Reduces oxidative stress and inflammatory responses.
  • Exhibits antiviral activity against HIV.
  • Inhibits urease in Helicobacter pylori and demonstrates antibacterial activity.
Catalog No. 50-004-12906 Supplier Medchemexpress LLC Supplier No. HYB123510MM/1ML
Acetohydroxamic acid is an inhibitor for bacterial and plant urease used for chronic urinary tract infections. It selectively inhibits arachidonic acid 5-lipoxygenase, useful in asthma research. This compound also inhibits advanced glycation end products, reduces oxidative stress and inflammatory responses, and exhibits antiviral activity against HIV.
  • Inhibitor for bacterial and plant urease.
  • Selectively inhibits arachidonic acid 5-lipoxygenase, applicable in asthma research.
  • Inhibits the formation of advanced glycation end products (AGEs).
  • Reduces oxidative stress and inflammatory responses.
  • Exhibits antiviral activity against HIV.
  • Inhibits urease in Helicobacter pylori and demonstrates antibacterial activity.