Learn More
Medchemexpress LLC Acetohydroxamic acid | 546-88-3 | 98.7% | 250 MG
SDP
Catalog No. 5000414409
Encompass_Preferred
Acetohydroxamic acid is an inhibitor for bacterial and plant urease, making it useful for chronic urinary tract infections. It selectively inhibits arachidonic acid 5-lipoxygenase, which is relevant for asthma research. The compound also inhibits the formation of advanced glycation end products and reduces oxidative stress and inflammatory responses. It has demonstrated antiviral activity against HIV.
Features and benefits:
  • Inhibits bacterial and plant urease
  • Selectively inhibits arachidonic acid 5-lipoxygenase
  • Reduces formation of advanced glycation end products
  • Reduces oxidative stress and inflammatory responses
  • Exhibits antiviral activity against HIV
Catalog No. 50-004-14409 Supplier Medchemexpress LLC Supplier No. 250MGHYB1235
Acetohydroxamic acid is an inhibitor for bacterial and plant urease, making it useful for chronic urinary tract infections. It selectively inhibits arachidonic acid 5-lipoxygenase, which is relevant for asthma research. The compound also inhibits the formation of advanced glycation end products and reduces oxidative stress and inflammatory responses. It has demonstrated antiviral activity against HIV.
Features and benefits:
  • Inhibits bacterial and plant urease
  • Selectively inhibits arachidonic acid 5-lipoxygenase
  • Reduces formation of advanced glycation end products
  • Reduces oxidative stress and inflammatory responses
  • Exhibits antiviral activity against HIV