Learn More
Medchemexpress LLC Acetohydroxamic acid | 546-88-3 | 98.7% | 500 MG
SDP
Catalog No. 5000412788
Encompass_Preferred
Acetohydroxamic acid (N-Hydroxyacetamide) is a chemical compound that acts as an inhibitor for bacterial and plant urease, commonly used for treating chronic urinary tract infections. It also exhibits selective inhibition of arachidonic acid 5-lipoxygenase, making it useful in asthma research. Beyond these, it inhibits the formation of advanced glycation end products and effectively reduces oxidative stress and inflammatory responses. This compound also demonstrates antiviral activity against HIV.
  • Inhibits bacterial and plant urease
  • Used for chronic urinary tract infections
  • Selectively inhibits arachidonic acid 5-lipoxygenase
  • Useful in asthma research
  • Inhibits advanced glycation end products formation
  • Reduces oxidative stress
  • Reduces inflammatory responses
  • Exhibits antiviral activity against HIV
Catalog No. 50-004-12788 Supplier Medchemexpress LLC Supplier No. HYB1235500MG
Acetohydroxamic acid (N-Hydroxyacetamide) is a chemical compound that acts as an inhibitor for bacterial and plant urease, commonly used for treating chronic urinary tract infections. It also exhibits selective inhibition of arachidonic acid 5-lipoxygenase, making it useful in asthma research. Beyond these, it inhibits the formation of advanced glycation end products and effectively reduces oxidative stress and inflammatory responses. This compound also demonstrates antiviral activity against HIV.
  • Inhibits bacterial and plant urease
  • Used for chronic urinary tract infections
  • Selectively inhibits arachidonic acid 5-lipoxygenase
  • Useful in asthma research
  • Inhibits advanced glycation end products formation
  • Reduces oxidative stress
  • Reduces inflammatory responses
  • Exhibits antiviral activity against HIV