Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acetoin

Catalog No. A17951250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A17951250G 250 g
A1795150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A17951250G Supplier Sigma Aldrich Supplier No. A17951250G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acetoin

Specifications

CAS 513-86-0
Melting Point 15°C (monomer), 90°C (dimer) (lit.)
Density 1.013 g/mL (at 25°C (literature))
Boiling Point 148°C (lit.)
Molecular Formula C4H8O2
Refractive Index n20/D 1.417 (literature)
Linear Formula CH3COCH(OH)CH3
MDL Number MFCD00004521
Quantity 250 g
Synonym 3-Hydroxy-2-butanone; Acetylmethylcarbinol
Molecular Weight (g/mol) 88.11
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.