Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetoin, MilliporeSigma™ Supelco™

Catalog No. 111016728 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6728 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6728 Supplier MilliporeSigma Supelco Supplier No. 40127U
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 513-86-0
Density 1.013 g/mL (at 25°C)
Flash Point 47°C
Physical Form Neat
Linear Formula CH3 COCH(OH)CH3
MDL Number MFCD00004521
Melting Point 15°C (monomer); 90°C (dimer)
Boiling Point 148°C
Synonym 3-Hydroxy-2-butanone; Acetylmethylcarbinol
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 88.11
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.