Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetoin primarily dimer 955KG
Catalog No. 502902759
Encompass
Acetoin is an acyloin derivative. In the presence of light and moisture it transforms into a white colored solid dimer. It is formed by the reduction of diacetyl. It has been synthesized from acetaldehyde.
Catalog No. 50-290-2759 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2008085KGK
Acetoin is an acyloin derivative. In the presence of light and moisture it transforms into a white colored solid dimer. It is formed by the reduction of diacetyl. It has been synthesized from acetaldehyde.