Learn More
Medchemexpress LLC Acetomenaphthone | 573-20-6 | 99.9% | 1 ML
SDP
Catalog No. 5000411956
Encompass_Preferred
Vitamin K4 is a chemically synthesized Vitamin K that plays an important role in the normal blood coagulation system. It arrests cells in the S phase and induces apoptosis. Vitamin K4 can be used for the research of cancer, such as prostate cancer and osteosarcoma.
  • Chemically synthesized Vitamin K
  • Plays an important role in the normal blood coagulation system
  • Arrests cells in S phase
  • Induces apoptosis
  • Used for research of cancer, such as prostate cancer and osteosarcoma
Catalog No. 50-004-11956 Supplier Medchemexpress LLC Supplier No. HYB150810MM/1ML
Vitamin K4 is a chemically synthesized Vitamin K that plays an important role in the normal blood coagulation system. It arrests cells in the S phase and induces apoptosis. Vitamin K4 can be used for the research of cancer, such as prostate cancer and osteosarcoma.
  • Chemically synthesized Vitamin K
  • Plays an important role in the normal blood coagulation system
  • Arrests cells in S phase
  • Induces apoptosis
  • Used for research of cancer, such as prostate cancer and osteosarcoma