Learn More
AdipoGen Acetomycin
SDP
Supplier:  AdipoGen BVT0150M005
Encompass_Preferred
Chemical. CAS 510-18-9. Formula C10H14O5. MW 214.2. Isolated from Streptomyces ramulosus. Antibiotic. Antibacterial, antifungal and antiprotozoal. Cytotoxic against several tumor cell lines in vitro.
Catalog No. 50-256-2087