Learn More
Electron Microscopy Sciences Acetone 1 QT
SDP
Catalog No. 5031733
Encompass_Preferred
Used in Alizarin Red S for calcium deposits staining. See references: McGee-Russell, S.M., J. Histochem., 6:22-42, 1958 or Clark, G., (ed), Staining Procedures, 3rd d., Williams & Wilkins, Baltimore, p. 140, c. 1973.
Catalog No. 50-317-33 Supplier Electron Microscopy Sciences Supplier No. 2620603
 Restricted Item
Add to Cart
Add to Cart
Used in Alizarin Red S for calcium deposits staining. See references: McGee-Russell, S.M., J. Histochem., 6:22-42, 1958 or Clark, G., (ed), Staining Procedures, 3rd d., Williams & Wilkins, Baltimore, p. 140, c. 1973.