Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone ACS reagent, >=99.5% | 67-64-1 | MFCD00008765 | 2.5L
Catalog No. NC2117493
Encompass
Acetone ACS reagent, >=99.5% | Purity: >=99.5% | Mol Wt: 58.08 | 67-64-1 | MFCD00008765 | 2.5L
This item is not returnable. View return policy
Catalog No. NC2117493 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1791242.5L
Acetone ACS reagent, >=99.5% | Purity: >=99.5% | Mol Wt: 58.08 | 67-64-1 | MFCD00008765 | 2.5L