Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetone, ACS Grade
SCI_ED

Catalog No. s25120 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25120 500 mL
S25120A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25120
Only null left
Add to Cart
Add to Cart

ACS grade solution for educational use. Ideal for analytical testing, laboratory procedures, and scientific research in classrooms and labs.

  • ACS Grade Solution
  • Product is for educational use
  • Available in 500 mL & 4 L sizes
  • Wide range of applications: analytical testing, lab procedures, and scientific research

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Density 0.790
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 100% v/v
Linear Formula CH3COCH3
Formula Weight 58.08
Grade Reagent
Chemical Name or Material Acetone
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.