Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone, 67-64-1, MFCD00008765, 4 L
Catalog No. NC0918592
Encompass
Linear Formula: CH3COCH3, Molecular Weight: 58.08, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%. Acetone is a ketone.
Catalog No. NC0918592 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6505014L
Linear Formula: CH3COCH3, Molecular Weight: 58.08, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%. Acetone is a ketone.