Learn More
Cambridge Isotope Laboratories Acetone-D6 (D, 99.9%) +0.03% V/V Tms, 666-52-4, MFCD00044635, 25G
Catalog No. NC1458374
Encompass_Preferred
Chemical Formula: CD3COCD3, Molecular Weight: 64.1, Synonym: Hexadeuteroacetone. Acetone-d6 may be used as an internal standard to detect aldehydes and acetone in water by headspace-solid-phase microextraction and gas chromatography-mass spectrometry.
Catalog No. NC1458374 Supplier Cambridge Isotope Laboratories Supplier No. DLM9TC25
Chemical Formula: CD3COCD3, Molecular Weight: 64.1, Synonym: Hexadeuteroacetone. Acetone-d6 may be used as an internal standard to detect aldehydes and acetone in water by headspace-solid-phase microextraction and gas chromatography-mass spectrometry.